Läs mer
Saccharin Sodium Salt Dihydrate, TRC
Produktkod. 30410766
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30410766 5 g
1 options
Produktkod. 30410766 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS0808105G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Saccharin Sodium Dihydrate
CAS 6155-57-3
Molekylformel C7 H4 N O3 S . Na . 2 H2 O
Kvantitet 5 g
Synonym 1,2-Benzisothiazol-3(2H)-one','1,1-dioxide','sodium salt','hydrate (1:1:2), 1,2-Benzisothiazol-3(2H)-one','1,1-dioxide','sodium salt','dihydrate (9CI), 1,2-Benzisothiazolin-3-one','1,1-dioxide','sodium salt','dihydrate (8CI), Saccharin sodium dihydrate, Sodium saccharin dihydrate, GR 50718C
InChI formel InChI=1S/C7H5NO3S.Na.2H2O/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;;;/h1-4H,(H,8,9);;2*1H2/q;+1;;/p-1
LEDER O.O.[Na+].O=C1[N-]S(=O)(=O)c2ccccc12
IUPAC-namn sodium;1,1-dioxo-1,2-benzothiazol-2-id-3-one;dihydrate
Molekylvikt (g/mol) 241.2
Formel vikt 241.0021