missing translation for 'onlineSavingsMsg'
Läs mer

Saclofen, TRC

Produktkod. 30419228
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419228 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419228 Supplier Toronto Research Chemicals Supplier No. TRCS08085010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Saclofen
CAS 125464-42-8
Molekylformel C9 H12 Cl N O3 S
Kvantitet 10 mg
Synonym Saclofen, β-(Aminomethyl)-4-chlorobenzeneethanesulfonic Acid
InChI formel InChI=1S/C9H12ClNO3S/c10-9-3-1-7(2-4-9)8(5-11)6-15(12,13)14/h1-4,8H,5-6,11H2,(H,12,13,14)
LEDER NCC(CS(=O)(=O)O)c1ccc(Cl)cc1
IUPAC-namn 3-amino-2-(4-chlorophenyl)propane-1-sulfonic acid
Molekylvikt (g/mol) 249.714
Formel vikt 249.023
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.