missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SACS Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22265-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ANKZF1 (ankyrin repeat and zinc finger domain containing 1), also known as ZNF744, is 726 amino acids in length and lacks a KRAB domain but contains one C2H2-type zinc finger and two ANK repeats. The gene encoding ANKZF1 localizes to chromosome 2.Specifications
SACS | |
Polyclonal | |
Unconjugated | |
SACS | |
ARSACS, DNAJC29, KIAA0730, SACS, Sacsin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26278, 50720 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JLC8, Q9NZJ4 | |
Sacs | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |