missing translation for 'onlineSavingsMsg'
Läs mer

Sacubitril Calcium, TRC

Produktkod. 30004554
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004554 100 mg
30004839 250 mg
30009306 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30004554 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS080900100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1369773-39-6
Molekylformel C24H28NO-5 1/2Ca2+
Molekylvikt (g/mol) 430.52
Synonym [S-(R*,S*)]-γ-[(3-Carboxy-1-oxopropyl)amino]-α-methyl-,[1,1'-biphenyl]-4-pentanoic Acid Ethyl Ester Calcium salt, (2R,4S)-4-[(3-Carboxy-1-oxopropyl)amino]-4-[(p-phenylphenyl)methyl]-2-methylbutanoic acid ethyl ester Calcium salt, (2R,4S)-5-(Biphenyl-4-yl)-4-[(3-carboxypropionyl)amino]-2-methylpentanoic acid ethyl ester Calcium salt, AHU 377 Calcium salt
IUPAC-namn calcium;4-[[(2S,4R)-5-ethoxy-4-methyl-5-oxo-1-(4-phenylphenyl)pentan-2-yl]amino]-4-oxobutanoate
LEDER C([C@H](C[C@H](C(OCC)=O)C)NC(CCC(O)=O)=O)C1=CC=C(C=C1)C2=CC=CC=C2.[Ca]

Specifications

Kemiskt namn eller material Sacubitril Calcium
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/2C24H29NO5.Ca/c2*1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19;/h2*4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28);/q;;+2/p-2/t2*17-,21+;/m11./s1
Formel vikt 860.3561
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.