Läs mer
Sacubitril Calcium, TRC
Produktkod. 30009306
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009306 1 g
30004554 100 mg
30004839 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009306 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS0809001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1369773-39-6
Molekylformel C24H28NO-5 1/2Ca2+
Molekylvikt (g/mol) 430.52
Synonym [S-(R*,S*)]-γ-[(3-Carboxy-1-oxopropyl)amino]-α-methyl-,[1,1'-biphenyl]-4-pentanoic Acid Ethyl Ester Calcium salt, (2R,4S)-4-[(3-Carboxy-1-oxopropyl)amino]-4-[(p-phenylphenyl)methyl]-2-methylbutanoic acid ethyl ester Calcium salt, (2R,4S)-5-(Biphenyl-4-yl)-4-[(3-carboxypropionyl)amino]-2-methylpentanoic acid ethyl ester Calcium salt, AHU 377 Calcium salt
IUPAC-namn calcium;4-[[(2S,4R)-5-ethoxy-4-methyl-5-oxo-1-(4-phenylphenyl)pentan-2-yl]amino]-4-oxobutanoate
LEDER C([C@H](C[C@H](C(OCC)=O)C)NC(CCC(O)=O)=O)C1=CC=C(C=C1)C2=CC=CC=C2.[Ca]
Specifikationer
Kemiskt namn eller material Sacubitril Calcium
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/2C24H29NO5.Ca/c2*1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19;/h2*4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28);/q;;+2/p-2/t2*17-,21+;/m11./s1
Formel vikt 860.3561
Procent renhet >95%