Beskrivning
- Folded 3-panel design adapts to various face shapes and sizes
- Sealing foams on both the upper and lower panels ensure an ideal seal while providing unmatched comfort for the wearer
- Easy to put on and adjust thanks to sliding elastic straps and a tightening ring
- The exhalation valve effectively expels heat and humidity, reducing the risk of fogging on the wearer's glasses
- Its individual packaging helps control the risk of contamination
Specifikationer
Specifikationer
| Färg | White, Blue |
| Designtyp | Flat-fold |
| Disponibel | Single-use |
| Filtertyp | FFP2 |
| Material | Polypropylene |
| Produkttyp | General Purpose Particulate Respirator |
| Storlek | One Size |
| Med utandningsventil | Yes |