Learn More
Brady™ Safety Sign COVID-19 General Safety Information
An efficient solution with clear safety instructions to easily communicate COVID-19 countermeasures on work floors.
48.25 SEK - 223.00 SEK
Specifications
Material | Laminated Polyester |
---|---|
Mounting Type | Self-adhesive |
Dimensions (L x W) | 371 x 262 mm (W x H) |
Product Code | Brand | Languages | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Languages | Price | Quantity & Availability | |||||
16506414
|
Brady™
306819 |
German |
223.00 SEK
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
16516414
|
Brady™
306820 |
English |
223.00 SEK
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
16526414
|
Brady™
306821 |
Spanish |
193.00 SEK
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16526434
|
Brady™
306841 |
Danish |
48.25 SEK
Each |
In Stock
Log in to see stock. |
|||||
16536414
|
Brady™
306822 |
Italian |
211.00 SEK
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16536434
|
Brady™
306842 |
Finnish |
48.25 SEK
Each |
In Stock
Log in to see stock. |
|||||
16536444
|
Brady™
306852 |
Norwegian |
48.25 SEK
Each |
In Stock
Log in to see stock. |
|||||
16546434
|
Brady™
306843 |
Swedish |
193.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
16586404
|
Brady™
306817 |
Dutch |
203.00 SEK
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16596404
|
Brady™
306818 |
French |
223.00 SEK
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
Description
- Provides clear, visual communication where it matters
- Highlights the use of appropriate personal protection
- Easily communicates social distancing and hygiene
Specifications
Laminated Polyester | |
Self-adhesive | |
371 x 262 mm (W x H) |