Learn More
Brady™ Safety Sign without Text - No Access for Unauthorized Persons
Product Code. 16200051 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
16200051 200 mm Each
15604207 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16200051 Supplier Brady™ Supplier No. 255016
  • ONLINE EXCLUSIVE PRICE
    175.00 SEK
    149.95 SEK / Paket med 1
    Save 25.05 SEK
    14% Av
View all promotions
Beräknad leverans: 27-11-2026
Safety sign for indicating mandatory action.
  • Improves workplace safety
Specifications
Alternate Part Number PIC 209-DIA 200-B7525
Material Aluminum
Diameter (Metric) 200 mm
Shape Round
Color Red on White