missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Q-Classic-30 Safety Storage Cabinet

For unrestricted storage of flammable hazardous materials in work areas.

Brand:  Asecos™ 31003-061-30005

Additional Details : Weight : 222.00000kg

 View more versions of this product

Product Code. 15231045

  • 18627.34 SEK / Each


Call for latest availability.
+46 (0)31 352 32 30

Add to basket

Description

Description

  • Tall cabinet with wing door
  • With cylinder locking and locking state indicator
  • Fire resistance of 30 minutes
  • In accordance with EN 14470-1 and TRGS 510
  • Tested in accordance with FM test standard 6050

Instrument & Equipment Services

10 years

Specifications

Specifications

EN 14470-1, DIN EN 14727, GS certified, CE compliant
Body: Antharcite grey, Door: Light grey RAL 7035; Sheet Steel RAL 7035
Winged
3X shelf, 1X bottom collecting sump (33L)
Powder-coated
1164mm
Steel
620mm
519 x 1050 x 1626mm
1947mm
Q30.195.116
Safety Storage Cabinet; Tall
Gray
Videos
Documentation