missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Q-Classic-90 Safety Storage Cabinet

For unrestricted storage of flammable hazardous materials in work areas.

Brand:  Asecos™ 30001-040-30005

Additional Details : Weight : 412.00000kg

 View more versions of this product

Product Code. 15271015

  • 27445.00 SEK / Each


Call for latest availability.
+46 (0)31 352 32 30

Add to basket

Description

Description

  • Robust safety storage for industrial sector
  • Tall cabinet with wing doors
  • With cylinder locking and locking state indicator
  • Integrated adjusting aids to compensate uneven floor
  • Sheet steel powder coated RAL 7035
  • Fire resistance of 90 minutes
  • In accordance with EN 14470-1 and TRGS 510
  • Tested in accordance with FM test standard 6050

Instrument & Equipment Services

10 years

Specifications

Specifications

EN 14470-1, DIN EN 14727, GS certified, CE compliant
Body: Anthracite grey RAL 7016, Door: Warning yellow RAL 1004; Sheet Steel RAL 7035
Winged
3X shelf, 1X bottom collecting sump (33L)
Powder-coated
1193mm
Steel
615mm
522 x 1050 x 1647mm
1953mm
Q-CLASSIC-90
Safety Storage Cabinet
Gray, Yellow
Videos