Läs mer
Saikosaponin D, TRC
Produktkod. 30421003
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421003 25 mg
30418942 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30421003 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS08391525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20874-52-6
Molekylformel C42 H68 O13
Molekylvikt (g/mol) 780.98
Synonym β-D-Galactopyranoside, (3β,4α,16α)-13,28-epoxy-16,23-dihydroxyolean-11-en-3-yl 6-deoxy-3-O-β-D-glucopyranosyl-, Saikosaponin D (8CI), (3β,4α,16α)-13,28-Epoxy-16,23-dihydroxyolean-11-en-3-yl 6-deoxy-3-O-β-D-glucopyranosyl-β-D-galactopyranoside
LEDER C[C@H]1O[C@@H](O[C@H]2CC[C@@]3(C)[C@@H](CC[C@]4(C)[C@@H]3C=C[C@]56OC[C@@]7(CCC(C)(C)C[C@@H]57)[C@H](O)C[C@@]46C)[C@]2(C)CO)[C@H](O)[C@@H](O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)[C@H]1O
Specifikationer
Kemiskt namn eller material Saikosaponin D
Kvantitet 25 mg
InChI formel InChI=1S/C42H68O13/c1-21-28(46)33(55-34-31(49)30(48)29(47)22(18-43)53-34)32(50)35(52-21)54-27-10-11-37(4)23(38(27,5)19-44)8-12-39(6)24(37)9-13-42-25-16-36(2,3)14-15-41(25,20-51-42)26(45)17-40(39,42)7/h9,13,21-35,43-50H,8,10-12,14-20H2,1-7H3/t21-,22-,23-,24-,25-,26-,27+,28+,29-,30+,31-,32-,33+,34+,35+,37+,38+,39-,40+,41-,42+/m1/s1
Formel vikt 780.466