Läs mer
Saint-Gobain Bio-Simplex™ Polypropylene, Carboy with Spigot
Produktkod. 16556660
:
Closed cap: 120mm; With spigot; 20L
Produktkod. 16556660
Brand: Saint-Gobain DCB20120S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
For Use With: Liquid handling and solution storage for general laboratory, process development and bioproduction applications; Autoclavable
TRUSTED_SUSTAINABILITY
Specifikationer
Noggrannhet ±5%
Autoklaveringsbar Autoclavable
Kapacitet (engelska) 5.3 gal.
Kapacitet (metrisk) 20 L
Certifikat och efterlevnad 21 CFR 177, USP Class VI
För användning med (applikation) Liquid handling and solution storage for general laboratory, process development and bioproduction applications
Inkluderar Bottom handles for full hand grip, closed cap and spigot
Material Polypropylene
Mun stil Wide
Produkttyp Carboy with Spigot