Learn More
Saint-Gobain™ Bytac™ Surface Protectors, 15L ft. x 25W in.; Backing: Aluminum
Brand: Saint-Gobain™ D1069329
Additional Details : Weight : 0.80000kg
Description
Put the protection of FEP fluoropolymer on any surface
Order Info
Bytac protector with vinyl backing recommended for temperatures up to 93°C (200°F); with aluminum foil backing, can be used up to 182°C (360°F).