Learn More
Saint-Gobain Norprene™ Pressure Tubing
7234.86 SEK - 10340.00 SEK
Specifications
Material Norprene
Overall Length (English) 50 ft.
View More Specs
Products 3
Product Code Brand Diameter (English) Inner Price Quantity & Availability  
Product Code Brand Diameter (English) Inner Price Quantity & Availability  
11882120
View Documents Promotion Details
Saint-Gobain
95715-02
1/2 in.
10340.00 SEK
15m
View alternative products
Save Up to 
»
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
11892120
View Documents Promotion Details
Saint-Gobain
95715-03
5/8 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11802130
View Documents Promotion Details
Saint-Gobain
95715-04
3/4 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications
Norprene
50 ft.