Learn More
Saint-Gobain Norprene™ Pressure Tubing
Brand:  Saint-Gobain 95715-04
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11802130
  • 7093.00 SEK / 15m
Estimated Shipment
23-06-2021
Specifications
85 psi
Norprene
50 ft.
3/4 in.
1.0625 in.