Läs mer
Salbutamol Acetonide, TRC
Produktkod. 30421737
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421737 500 mg
30415902 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30421737 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS085530500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54208-72-9
Molekylformel C16 H25 N O3
Molekylvikt (g/mol) 279.37
Synonym α-[[(1,1-Dimethylethyl)amino]methyl]-2,2-dimethyl-4H-1,3-benzodioxin-6-methanol, 2-(tert-Butylamino)-1-(2,2-dimethyl-1,3-benzodioxin-6-yl)ethanol, Salbutamol Acetonide
IUPAC-namn 2-(tert-butylamino)-1-(2,2-dimethyl-4H-1,3-benzodioxin-6-yl)ethanol
LEDER CC(C)(C)NCC(O)c1ccc2OC(C)(C)OCc2c1
Specifikationer
Kemiskt namn eller material Salbutamol Acetonide
Kvantitet 500 mg
InChI formel InChI=1S/C16H25NO3/c1-15(2,3)17-9-13(18)11-6-7-14-12(8-11)10-19-16(4,5)20-14/h6-8,13,17-18H,9-10H2,1-5H3
Formel vikt 279.1834