Läs mer
Salbutamon Hydrochloride, TRC
Produktkod. 30426846
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30426846 10 mg -
30420717 25 mg -
30420527 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426846 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS08550010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41489-89-8
Molekylformel C13 H19 N O3 . Cl H
Molekylvikt (g/mol) 273.76
Synonym 2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride, Salbutamol Sulphate Imp. J (EP) as Hydrochloride, Salbutamone Hydrochloride
IUPAC-namn 2-(tert-butylamino)-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone;hydrochloride
LEDER Cl.CC(C)(C)NCC(=O)c1ccc(O)c(CO)c1
Specifikationer
Kemiskt namn eller material 2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride (Salbutamone Hydrochloride)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C13H19NO3.ClH/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;/h4-6,14-16H,7-8H2,1-3H3;1H
Formel vikt 273.1132
Procent renhet >95%