Läs mer
Salicylazoiminopyridine (>85%), TRC
Produktkod. 30423411
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423411 250 mg
30415905 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423411 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS088100250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66030-25-9
Molekylformel C18H14N4O3
Molekylvikt (g/mol) 334.33
Synonym 2-Hydroxy-5-[2-[4-(2-imino-1(2H)-pyridinyl)phenyl]diazenyl]benzoic Acid, 2-Hydroxy-5-[[4-(2-imino-1(2H)-pyridinyl)phenyl]azo]benzoic Acid, Sulfasalazine Impurity C
IUPAC-namn 2-hydroxy-5-[[4-(2-iminopyridin-1-yl)phenyl]diazenyl]benzoic acid
LEDER OC(=O)c1cc(ccc1O)N=Nc2ccc(cc2)N3C=CC=CC3=N
Specifikationer
Kemiskt namn eller material Salicylazoiminopyridine
Kvantitet 250 mg
InChI formel InChI=1S/C18H14N4O3/c19-17-3-1-2-10-22(17)14-7-4-12(5-8-14)20-21-13-6-9-16(23)15(11-13)18(24)25/h1-11,19,23H,(H,24,25)
Formel vikt 334.1066