Läs mer
Salicylic Acid beta-D-O-Glucuronide, TRC
Produktkod. 30135557
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135557 50 mg
30005004 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135557 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS08813050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7695-70-7
Molekylformel C13 H14 O9
Molekylvikt (g/mol) 314.24
Synonym β-D-Glucopyranosiduronic acid, 2-carboxyphenyl, Glucopyranosiduronic acid, o-carboxyphenyl, β-D- (8CI), β-D-Glucopyranosiduronic acid, o-carboxyphenyl (6CI,7CI), 2-Carboxyphenyl β-D-glucopyranosiduronic acid, Salicyl glucuronide, Salicyl phenolic glucuronide, Salicylic acid glucuronide, Salicylic acid phenolic glucuronide, Salicylic glucuronide
IUPAC-namn (2S,3S,4S,5R,6S)-6-(2-carboxyphenoxy)-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER O[C@@H]1[C@@H](O)[C@H](Oc2ccccc2C(=O)O)O[C@@H]([C@H]1O)C(=O)O
Specifikationer
Kemiskt namn eller material Salicylic Acid beta-D-O-Glucuronide
Kvantitet 50 mg
InChI formel InChI=1S/C13H14O9/c14-7-8(15)10(12(19)20)22-13(9(7)16)21-6-4-2-1-3-5(6)11(17)18/h1-4,7-10,13-16H,(H,17,18)(H,19,20)/t7-,8-,9+,10-,13+/m0/s1
Formel vikt 314.0638