missing translation for 'onlineSavingsMsg'
Läs mer

Salicylic acid-pentyl ester 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Produktkod. 30309535
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30309535 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30309535 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA16904200AL1000

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Salicylic acid-pentyl ester
CAS 2050-08-0
Analyt- eller komponentnamn Salicylic acid-pentyl ester
Frakt skick +20°C
Molekylformel C12 H16 O3
Koncentration 1000 μg/mL
Certifikat och efterlevnad ISO 17034
Synonym Salicylic acid, amyl ester (4CI), Salicylic acid, pentyl ester (6CI,8CI), 2-Hydroxybenzoic acid pentyl ester, Amyl salicylate, NSC 403668, NSC 44877, NSC 46125, Pentyl salicylate
InChI formel InChI=1S/C12H16O3/c1-2-3-6-9-15-12(14)10-7-4-5-8-11(10)13/h4-5,7-8,13H,2-3,6,9H2,1H3
LEDER CCCCCOC(=O)c1ccccc1O
IUPAC-namn pentyl 2-hydroxybenzoate
Lösningsmedel eller matris Acetonitrile
Molekylvikt (g/mol) 208.25
Kvantitet 1 ml
Formel vikt 208.1099 g/mol
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.