Läs mer
Salinomycin Sodium Salt (12% min.), TRC
Produktkod. 30138213
Change view
Kvantitet:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138213 1 g
30132421 10 g
30005097 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138213 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS0888001G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55721-31-8
Molekylformel C42 H69 O11 . Na
Molekylvikt (g/mol) 772.98
Synonym Salinomycin Sodium, Salinomycin, monosodium salt (9CI), 1,6,8-Trioxadispiro[4.1.5.3]pentadecane, salinomycin deriv., Kokcisan, Kokcisan 120G, Sacox, Salinomycin sodium, Salinomycin sodium salt, Salinopharm-premix, Sodium salinomycin, Synvertas plv. a.u.v., Salinomycin, sodium salt (1:1)
IUPAC-namn sodium;(2R)-2-[(2R,5S,6R)-6-[(2S,3S,4S,6R)-6-[(3S,5S,7R,9S,10S,12R,15R)-3-[(2R,5R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-3,10,12-trimethyl-4,6,8-trioxadispiro[4.1.5^{7}.3^{5}]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-oxooctan-2-yl]-5-methyloxan-2-yl]butanoate
LEDER [Na+].CC[C@H]([C@H]1CC[C@H](C)[C@@H](O1)[C@@H](C)[C@H](O)[C@H](C)C(=O)[C@H](CC)[C@H]2O[C@@]3(O[C@@]4(CC[C@](C)(O4)[C@H]5CC[C@](O)(CC)[C@H](C)O5)[C@H](O)C=C3)[C@H](C)C[C@@H]2C)C(=O)[O-]
Specifikationer
Kemiskt namn eller material Salinomycin sodium salt
Kvantitet 1 g
InChI formel InChI=1S/C42H70O11.Na/c1-11-29(38(46)47)31-15-14-23(4)36(50-31)27(8)34(44)26(7)35(45)30(12-2)37-24(5)22-25(6)41(51-37)19-16-32(43)42(53-41)21-20-39(10,52-42)33-17-18-40(48,13-3)28(9)49-33;/h16,19,23-34,36-37,43-44,48H,11-15,17-18,20-22H2,1-10H3,(H,46,47);/q;+1/p-1/t23-,24-,25+,26-,27-,28-,29+,30-,31+,32+,33+,34+,36+,37-,39-,40+,41-,42-;/m0./s1
Formel vikt 772.4738