Läs mer
Salinomycin Sodium Salt, TRC
Produktkod. 30007833
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007833 250 mg
18093020 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007833 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS088805250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55721-31-8
Molekylformel C42 H69 O11 . Na
Molekylvikt (g/mol) 772.98
Synonym Salinomycin Sodium, Salinomycin, monosodium salt (9CI), 1,6,8-Trioxadispiro[4.1.5.3]pentadecane, salinomycin deriv., Kokcisan, Kokcisan 120G, Sacox, Salinomycin sodium, Salinomycin sodium salt, Salinopharm-premix, Sodium salinomycin, Synvertas plv. a.u.v., Salinomycin, sodium salt (1:1)
IUPAC-namn sodium;(2R)-2-[(2R,5S,6R)-6-[(2S,3S,4S,6R)-6-[(3S,5S,7R,9S,10S,12R,15R)-3-[(2R,5R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-3,10,12-trimethyl-4,6,8-trioxadispiro[4.1.5^{7}.3^{5}]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-oxooctan-2-yl]-5-methyloxan-2-yl]butanoate
LEDER [Na+].CC[C@H]([C@H]1CC[C@H](C)[C@@H](O1)[C@@H](C)[C@H](O)[C@H](C)C(=O)[C@H](CC)[C@H]2O[C@@]3(O[C@@]4(CC[C@](C)(O4)[C@H]5CC[C@](O)(CC)[C@H](C)O5)[C@H](O)C=C3)[C@H](C)C[C@@H]2C)C(=O)[O-]
Specifikationer
Kemiskt namn eller material Salinomycin sodium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C42H70O11.Na/c1-11-29(38(46)47)31-15-14-23(4)36(50-31)27(8)34(44)26(7)35(45)30(12-2)37-24(5)22-25(6)41(51-37)19-16-32(43)42(53-41)21-20-39(10,52-42)33-17-18-40(48,13-3)28(9)49-33;/h16,19,23-34,36-37,43-44,48H,11-15,17-18,20-22H2,1-10H3,(H,46,47);/q;+1/p-1/t23-,24-,25+,26-,27-,28-,29+,30-,31+,32+,33+,34+,36+,37-,39-,40+,41-,42-;/m0./s1
Formel vikt 772.4738
Procent renhet >95%