missing translation for 'onlineSavingsMsg'
Learn More

Salvianolic acid C, MedChemExpress

Product Code. p-200096050
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Kvantitet:
5 mg
This item is not returnable. View return policy

Product Code. 16471378

missing translation for 'mfr': Medchem Express HYN0319

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

MedChemExpress Salvianolic acid C is a noncompetitive Cytochrome P4502C8 (CYP2C8) inhibitor and a moderate mixed inhibitor of Cytochrome P45022J2 (CYP2J2), with Kis of 4.82 μM and 5.75 μM for CYP2C8 and CYP2J2, respectively.

Salvianolic acid C is a noncompetitive Cytochrome P4502C8 (CYP2C8) inhibitor and a moderate mixed inhibitor of Cytochrome P45022J2 (CYP2J2), with Kis of 4.82 μM and 5.75 μM for CYP2C8 and CYP2J2, respectively.

Specifications

Kemiskt namn eller material Salvianolic acid C
CAS 115841-09-3
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C26H20O10
Kvantitet 5 mg
Löslighetsinformation DMSO : 50 mg/mL (101.54 mM; Need ultrasonic)
LEDER OC1=C(O)C=CC(C[C@H](C(O)=O)OC(/C=C/C2=CC=C(O)C3=C2C=C(C4=CC(O)=C(O)C=C4)O3)=O)=C1
Molekylvikt (g/mol) 492.43
Formel vikt 492.43
Procent renhet 98.51%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.