Läs mer
Salvinorin A, Lipomed
Produktkod. 30459244
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30459244 100 mg
1 options
Produktkod. 30459244 Leverantör Lipomed Leverantörsnummer LPMSLV1095100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Salvinorin A
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Salvinorin A
CAS 83729-01-5
Analyt- eller komponentnamn Salvinorin A
Frakt skick Room Temperature
Molekylformel C23 H28 O8
Synonym 2 H-Naphtho[2,1-c]pyran-7-carboxylic acid, 9-(acetyloxy)-2-(3-furanyl)dodecahydro-6 A,10 b-dimethyl-4,10-dioxo-, methyl ester, (2 S,4 aR,6 aR,7 R,9 S,10 AS,10 bR)-, 2 H-Naphtho[2,1-c]pyran-7-carboxylic acid, 9-(acetyloxy)-2-(3-furanyl)dodecahydro-6 A,10 b-dimethyl-4,10-dioxo-, methyl ester, [2 S-(2α,4 Aα,6 Aβ,7β,9β,10 Aα,10 bβ)]-, (-)-Salvinorin A, Divinorin A, Salvinorin, Salvinorin A
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER COC(=O)[C@@H]1 C[C@H](OC(=O)C)C(=O)[C@H]2 [C@@]1(C)CC[C@H]3 C(=O)O[C@@H](C[C@]23 C)c4cocc4
IUPAC-namn methyl (2 S,4 aR,6 aR,7 R,9 S,10 AS,10 bR)-9-acetyloxy-2-(furan-3-yl)-6 A,10 b-dimethyl-4,10-dioxo-2,4 A,5,6,7,8,9,10 A-octahydro-1 H-benzo[f]isochromene-7-carboxylate
Industry Type Pharmaceutical
Molekylvikt (g/mol) 432.46
Kvantitet 100 mg
Formel vikt 432.1784 g/mol