Learn More
Sam50 Mouse anti-Human, Unconjugated, Clone: 2H6H9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67425-1-IG-20UL
Description
The Sam50 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Sam50. This antibody recognizes human, pig, rat, mouse antigen. The Sam50 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Sam50 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2H6H9 | |
Unconjugated | |
20 μL | |
25813 | |
IgG1 |