missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26815-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.Specifications
SAMD8 | |
Polyclonal | |
Unconjugated | |
SAMD8 | |
SAMD8, SMSr | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
142891 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96LT4 | |
SAMD8 | |
SAMD8 Fusion Protein Ag25310 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |