Learn More
Masterflex™ Sterilized Sampling Bottle Assemblies
Product Code. 13286950 Shop All Masterflex Products
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
13286950 500 mL Each
1 options
Product Code. 13286950 Supplier Masterflex™ Supplier No. 9564404
  • ONLINE EXCLUSIVE PRICE
    1660.00 SEK
    1514.66 SEK / Paket med 1
    Save 145.34 SEK
    8% Av
Beräknad leverans: 04-01-2027
Transfer, sample and store biopharmaceutical products and reagents in applications from research to process with these sample bottles. Cole-Parmer™ Sterilized Sampling Bottle Assemblies are pre-sterilized by gamma irradiation.
Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections.
Specifications
Capacity (Metric) 500 mL
For Use With (Application) Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to process.
Material Plastic
Closure Included Yes
Closure Liner Seamless