Läs mer
Sanguinarium Chloride, TRC
Produktkod. 30007873
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007873 10 mg
30004483 50 mg
30005083 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007873 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS11250010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5578-73-4
Molekylformel C20 H14 N O4 . Cl
Molekylvikt (g/mol) 367.78
Synonym [1,3]Benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridinium, 13-methyl-, chloride (9CI), Sanguinarine, chloride (6CI, 8CI), 13-Methyl[1,3]benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridinium chloride, Sanguinarine hydrochloride, Sanguinarium chloride
LEDER [Cl-].C[n+]1cc2c3OCOc3ccc2c4ccc5cc6OCOc6cc5c14
Specifikationer
Kemiskt namn eller material Sanguinarine chloride
Kvantitet 10 mg
InChI formel InChI=1S/C20H14NO4.ClH/c1-21-8-15-12(4-5-16-20(15)25-10-22-16)13-3-2-11-6-17-18(24-9-23-17)7-14(11)19(13)21;/h2-8H,9-10H2,1H3;1H/q+1;/p-1
Formel vikt 367.0611