missing translation for 'onlineSavingsMsg'
Läs mer

Santacruzamate A, MedChemExpress

Produktkod. 16431618
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16431618 5 mg -
16429208 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16431618 Leverantör Medchem Express Leverantörsnummer HYN0931

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Santacruzamate A (CAY-10683) is a potent and selective HDAC2 inhibitor with an IC50 of 119 pM.

Santacruzamate A (CAY-10683) is a potent and selective HDAC2 inhibitor with an IC50 of 119 pM.

Specifikationer

Kemiskt namn eller material Santacruzamate A
CAS 1477949-42-0
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C15H22N2O3
Kvantitet 5 mg
Synonym CAY-10683
Löslighetsinformation DMSO : ≥ 100 mg/mL (359.26 mM)
LEDER O=C(OCC)NCCCC(NCCC1=CC=CC=C1)=O
Molekylvikt (g/mol) 278.35
Formel vikt 278.35
Procent renhet 99.32%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death
Visa mer Visa mindre

Research purposes only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.