Läs mer
Santamarine, TRC
Produktkod. 30005480
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30005480 25 mg -
30008342 2.5 mg 2.5mg
30005669 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005480 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS12423525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4290-13-5
Molekylformel C15H20O3
Molekylvikt (g/mol) 248.32
Synonym (3aS,5aR,6R,9aS,9bS)-3a,4,5,5a,6,7,9a,9b-Octahydro-6-hydroxy-5a,9-dimethyl-3-methylenenaphtho[1,2-b]furan-2(3H)-one Santamarin Balchanin
IUPAC-namn (3aS,5aR,6R,9aS,9bS)-6-hydroxy-5a,9-dimethyl-3-methylidene-4,5,6,7,9a,9b-hexahydro-3aH-benzo[g][1]benzofuran-2-one
LEDER CC1=CC[C@@H](O)[C@]2(C)CC[C@@H]3[C@H](OC(=O)C3=C)[C@@H]12
Specifikationer
Kemiskt namn eller material Santamarine
Kvantitet 25 mg
InChI formel InChI=1S/C15H20O3/c1-8-4-5-11(16)15(3)7-6-10-9(2)14(17)18-13(10)12(8)15/h4,10-13,16H,2,5-7H2,1,3H3/t10-,11+,12+,13-,15-/m0/s1
Formel vikt 248.1412