missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ SAPAP1 Polyclonal Antibody

Beskrivning
Additional Information: This antibody is expected to recognize both reported isoforms (NP_001003809.1; NP_004737.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Part of the postsynaptic scaffold in neuronal cells. Target binds the guanylate kinase-like domain of the PSD-95 family of synaptic ion channel clustering proteins; may play a role in protein-protein interactions at the synaptic junction.
Specifikationer
Specifikationer
| Antigen | SAPAP1 |
| Användningsområden | Immunohistochemistry (Paraffin) |
| Klassificering | Polyclonal |
| Koncentration | 0.5 mg/mL |
| Konjugera | Unconjugated |
| Formulering | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gen | Dlgap1 |
| Genaccessionsnr. | O14490 |
| Gene Alias | 4933422O14Rik; 9630002F18; AI845682; AI848168; BB075781; D17Bwg0511e; DAP1; DAP-1; DAP-1 beta; DAP-1-ALPHA; DAP-1-BETA; discs large homolog associated protein 1; discs, large (Drosophila) homolog-associated protein 1; disks large-associated protein 1; DLG associated protein 1; Dlgap1; DLGAP1A; DLGAP1B; GKAP; GKAP/SAPAP; GKAP/SAPAP1; GKPA/SAPAP; guanylate kinase associated protein; guanylate kinase-associated protein; hGKAP; Kiaa4162; mKIAA4162; PSD-95 binding protein; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-binding protein 1; rGKAP; SAP90/PSD-95 associated protein 1; SAP90/PSD-95-associated protein 1; SAPAP1 |
| Gensymboler | Dlgap1 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?