Läs mer
Tocris Bioscience™ Saquinavir mesylate
GREENER_CHOICE
Produktkod. 18095832
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18095832 50 mg
18075322 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18095832 Leverantör Tocris Bioscience™ Leverantörsnummer 4418/50
HIV protease inhibitor
Saquinavir mesylate is an inhibitor of human immunodeficiency virus (HIV) protease (Ki values are <0.1 and 0.12 nM for HIV-2 and HIV-1 protease respectively). Exhibits high antiviral activity and low cytotoxicity. In silico docking models predict potential as an inhibitor of SARS-CoV-2 3CLpro.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 149845-06-7
Molekylformel C39H54N6O8S
Specifikationer
Kemiskt namn eller material Saquinavir mesylate
Kvantitet 50 mg
Formel vikt 766.95
Renhet >99%
Molekylvikt (g/mol) 766.955