Läs mer
Saquinavir Mesylate, TRC
Produktkod. 30137846
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137846 50 mg
30134811 25 mg
30007108 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137846 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS13500050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 149845-06-7
Molekylformel C38 H50 N6 O5 . C H4 O3 S
Molekylvikt (g/mol) 766.95
Synonym Saquinavir mesilate, (2S)-N1-[(1S,2R)-1-Benzyl-3-[(3S,4aS,8aS)-3-[(1,1-dimethylethyl)carbamoyl]octahydroisoquinolin-2(1H)-yl]-2-hydroxypropyl]-2-[(quinolin-2-ylcarbonyl)amino]butanediamide methanesulfonate, Butanediamide, N1-[(1S,2R)-3-[(3S,4aS,8aS)-3-[[(1,1-dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]-, (2S)-, methanesulfonate (1:1), Butanediamide, N1-[(1S,2R)-3-[(3S,4aS,8aS)-3-[[(1,1-dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]-, (2S)-, monomethanesulfonate (salt) (9CI), Invirase, Ro 31-8959/003, Saquinavir mesylate, Butanediamide, N1-[3-[3-[[(1,1-dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]-, [3S-[2[1R*(R*),2S*],3alpha,4abeta,8abeta]]-, monomethanesulfonate (salt)
IUPAC-namn (2S)-N-[(2S,3R)-4-[(3S,4aS,8aS)-3-(tert-butylcarbamoyl)-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinolin-2-yl]-3-hydroxy-1-phenylbutan-2-yl]-2-(quinoline-2-carbonylamino)butanediamide;methanesulfonic acid
LEDER CC(C)(C)NC(=O)[C@@H]1C[C@@H]2CCCC[C@@H]2CN1C[C@@H](O)[C@H](Cc3ccccc3)NC(=O)[C@H](CC(=O)N)NC(=O)c4ccc5ccccc5n4.CS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Saquinavir mesilate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C38H50N6O5.CH4O3S/c1-38(2,3)43-37(49)32-20-26-14-7-8-15-27(26)22-44(32)23-33(45)30(19-24-11-5-4-6-12-24)41-36(48)31(21-34(39)46)42-35(47)29-18-17-25-13-9-10-16-28(25)40-29;1-5(2,3)4/h4-6,9-13,16-18,26-27,30-33,45H,7-8,14-15,19-23H2,1-3H3,(H2,39,46)(H,41,48)(H,42,47)(H,43,49);1H3,(H,2,3,4)/t26-,27+,30-,31-,32-,33+;/m0./s1
Formel vikt 766.3724
Procent renhet >95%