Läs mer
SAR405, MedChemExpress
Produktkod. 16470166
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16470166 2 mg
16455198 10 mg
16451417 50 mg
16433316 25 mg
16417927 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16470166 Leverantör Medchem Express Leverantörsnummer HY124812mg
MedChemExpress SAR405 is a first-in-class, selective, and ATP-competitive PI3K class III (PIK3C3) isoform Vps34 inhibitor (IC50=1.2 nM; Kd=1.5 nM). SAR405 inhibits autophagy induced either by starvation or by mTOR inhibition. Anticancer activity.
SAR405 is a first-in-class, selective, and ATP-competitive PI3K class III (PIK3C3) isoform Vps34 inhibitor (IC50=1.2 nM
Specifikationer
Kemiskt namn eller material SAR405
Fysisk form Solid
CAS 1523406-39-4
Färg Light Yellow
Kvantitet 2 mg
Anteckningar om renhetsgrad Research
Molekylformel C19H21ClF3N5O2
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : ≥ 27 mg/mL (60.83 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER O=C1N(C2=NC(N3[C@H](C)COCC3)=C1)CC[C@@H](C(F)(F)F)N2CC4=CC(Cl)=CN=C4
Molekylvikt (g/mol) 443.85
Formel vikt 443.85
Procent renhet 97.03%