missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
SAR407899, MedChemExpress
Beskrivning
SAR407899 is a selective, potent and ATP-competitive ROCK inhibitor, with an IC50 of 135 nM for ROCK-2, and Kis of 36 nM and 41 nM for human and rat ROCK-2, respectively.
Specifikationer
Specifikationer
| Kemiskt namn eller material | SAR407899 |
| CAS | 923359-38-0 |
| Färg | Light Yellow |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C14H16N2O2 |
| Kvantitet | 100 mg |
| Löslighetsinformation | DMSO : 6 mg/mL (24.56 mM; Need warming) |
| LEDER | O=C(NC=C1)C(C1=C2)=CC=C2OC3CCNCC3 |
| Molekylvikt (g/mol) | 244.29 |
| Formel vikt | 244.29 |
| Visa mer |
Research purposes only
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?