Läs mer
Saralasin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Saralasin |
| CAS | 34273-10-4 |
| Molekylformel | C42 H65 N13 O10 |
| Kvantitet | 50 mg |
| Synonym | Saralasin, L-Alanine','N-methylglycyl-L-arginyl-L-valyl-L-tyrosyl-L-valyl-L-histidyl-L-prolyl- (9CI','ACI), Angiotensin II','1-(N-methylglycine)-5-L-valine-8-L-alanine- (ZCI), Angiotensin II','1-sarcosine-5-L-valine-8-L-alanine- (8CI), N-Methylglycyl-L-arginyl-L-valyl-L-tyrosyl-L-valyl-L-histidyl-L-prolyl-L-alanine (ACI), 1-Sar-5-Val-8-Ala-angiotensin II, 1-Sar-8-ala-angiotensin II, [1-Sarcosine-5-valine-8-alanine]angiotensin II, Aralasin, P 113, P 113 (angiotensin II derivative), Saralasin |
| InChI formel | InChI=1S/C42H65N13O10/c1-22(2)33(53-35(58)28(50-32(57)20-45-6)9-7-15-47-42(43)44)38(61)51-29(17-25-11-13-27(56)14-12-25)36(59)54-34(23(3)4)39(62)52-30(18-26-19-46-21-48-26)40(63)55-16-8-10-31(55)37(60)49-24(5)41(64)65/h11-14,19,21-24,28-31,33-34,45,56H,7- |
| LEDER | CNCC(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](C)C(=O)O |
| IUPAC-namn | (2S)-2-[[(2S)-1-[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-5-carbamimidamido-2-[[2-(methylamino)acetyl]amino]pentanoyl]amino]-3-methylbutanoyl]amino]-3-(4-hydroxyphenyl)propanoyl]amino]-3-methylbutanoyl]amino]-3-(1H-imidazol-4-yl)propanoyl]pyrrolidine-2-ca |
| Molekylvikt (g/mol) | 912.047 |
| Formel vikt | 911.498 |