missing translation for 'onlineSavingsMsg'
Läs mer

Sarcalumenin Antibody, Novus Biologicals™

Produktkod. 18051738 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
25ul
Förpackningsstorlek:
0.1ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
18051738 0.1 mL 0.1ml
18422712 25ul -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18051738 Leverantör Novus Biologicals Leverantörsnummer NBP213378
  • ONLINE EXCLUSIVE PRICE
    5960.00 SEK
    5634.30 SEK / 0.1ml
    Save 325.70 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 17-09-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Sarcalumenin Polyclonal specifically detects Sarcalumenin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sarcalumenin
Användningsområden Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassificering Polyclonal
Konjugera Unconjugated
Utspädning Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulering PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias sarcalumenin
Gensymboler SRL
Värd art Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: ETEDANEEAPLRDRSHIEKTLMLNEDKPSDDYSAVLQRLRKIYHSSIKPLEQSYKYNELRQHEITDGEITSKPMVLFLGPWSVGKSTMINYLLGLENTRYQL
Reningsmetod Affinity Purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Apoptosis, Cancer, Tumor Suppressors
Primär eller sekundär Primary
Gen-ID (Entrez) 6345
Testspecificitet Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Målarter Human
Innehåll och lagring Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotyp IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.