missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ SARM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143637
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Specific for endogenous levels of the ~79 kDa SARM1 protein.
NAD(+) hydrolase, which plays a key role in axonal degeneration following injury by regulating NAD(+) metabolism (PubMed:25908823, PubMed:27671644, PubMed:28334607). Acts as a negative regulator of MYD88- and TRIF-dependent toll-like receptor signaling pathway by promoting Wallerian degeneration, an injury-induced form of programmed subcellular death which involves degeneration of an axon distal to the injury site (PubMed:15123841, PubMed:16964262, PubMed:20306472, PubMed:25908823). Wallerian degeneration is triggered by NAD(+) depletion: in response to injury, SARM1 is activated and catalyzes cleavage of NAD(+) into ADP-D-ribose (ADPR), cyclic ADPR (cADPR) and nicotinamide; NAD(+) cleavage promoting cytoskeletal degradation and axon destruction (PubMed:25908823, PubMed:28334607, PubMed:30333228, PubMed:31128467, PubMed:31439793, PubMed:31439792). Also able to hydrolyze NADP(+), but not other NAD(+)-related molecules (PubMed:29395922). Can activate neuronal cell death in response to stress (PubMed:20306472). Regulates dendritic arborization through the MAPK4-JNK pathway. Involved in innate immune response: inhibits both TICAM1/TRIF- and MYD88-dependent activation of JUN/AP-1, TRIF-dependent activation of NF-kappa-B and IRF3, and the phosphorylation of MAPK14/p38 (PubMed:16964262). [UniProt].
Specifikationer
| SARM | |
| Polyclonal | |
| Unconjugated | |
| SARM1 | |
| A830091I15Rik; C78606; FLJ36296; KIAA0524; MyD885; MyD88-5; SAM domain-containing protein 2; SAMD2; SARM; SARM1; Sterile alpha and Armadillo repeat protein; sterile alpha and HEAT/Armadillo motif containing 1; sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila; sterile alpha and TIR motif containing 1; sterile alpha and TIR motif-containing protein 1; Sterile alpha motif domain-containing protein 2; tir-1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23098, 237868, 287545 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| 10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5 | |
| Q6PDS3, Q6SZW1 | |
| SARM1 | |
| Full length recombinant human SARM1 protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering