Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Inhibitor screening Kit
This kit is useful for screening inhibitors of SARS-CoV-2.
Brand: ACROBiosystems EP-105-96tests
Additional Details : Weight : 0.00010kg
Description
The newly identified 2019 novel coronavirus (SARS-CoV-2) has posed a serious threat to human health. Currently, there is no specific treatment for COVID-19. It is urgent to develop the SARS-CoV-2 inhibitors, for example vaccines and therapeutic antibodies and small molecular compounds against COVID-19.Specifications
Inhibitor Screening Kits | |
96 Tests | |
-20°C, aviod light |
This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of SARS-CoV-2 inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized SARS-CoV-2 S protein RB | |
RT |