Läs mer
SARS-CoV-2 Envelope Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-07060MFV500
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SARS-CoV-2 Envelope Polyclonal antibody specifically detects SARS-CoV-2 Envelope in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifikationer
| Immunohistochemistry | |
| Western Blot, ELISA, Immunohistochemistry | |
| Rabbit | |
| 0.1 mL |
| Polyclonal | |
| 50mM Sodium Borate | |
| This antibody was raised against a peptide corresponding to 14 amino acids near the carboxyl terminus of SARS-CoV-2 (COVID-19) Envelope protein. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19) Envelope protein. | |
| Purified |