missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (N501Y), Fc Tag
5340.66 SEK - 38586.31 SEK
Specifications
Molecular Weight (g/mol) | 51.1 kDa |
---|---|
Source | SARS-CoV-2 S protein RBD (N501Y), Fc Tag (SPD-C5253) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)). |
Immunogen | Arg 319 - Lys 537 |
Storage Requirements | -20°C |
Gene Alias | Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17206510
|
ACROBiosystems
SPD-C5253-100UG |
100 μg |
5340.66 SEK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17216510
|
ACROBiosystems
SPD-C5253-1MG |
1 mg |
38586.31 SEK
1mg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.1 kDa.Specifications
51.1 kDa | |
Arg 319 - Lys 537 | |
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19 | |
Unconjugated | |
Fc Tag | |
Powder |
SARS-CoV-2 S protein RBD (N501Y), Fc Tag (SPD-C5253) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)). | |
-20°C | |
S protein RBD | |
SARS-CoV-2 | |
HEK293 | |
Viral Proteins |