missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
SARS-CoV-2 Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB105802-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Spike RBD Monoclonal specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifikationer
| SARS-CoV-2 Spike RBD | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein | |
| Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD Protein, Ala319-Phe541, Accession # YP_009724390.1 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry, Blocking Assay | |
| 1035423 | |
| Immunohistochemistry 8-25 ug/mL, Blockade of Receptor-ligand Interaction | |
| YP_009724390.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 43740568 | |
| SARS-CoV-2 | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering