Learn More
Sartorius Stedim Biotech™ Sartolab BT Vacuum Filter, 0.45 μm, Cellulose Acetate, 500 mL
Designed for particle and cell removal, this filter can be used for clarification purposes.
Brand:  Sartorius Stedim Biotech™ 180A4----------E
Additional Details : Weight : 1.36000kg
Product Code. 15982590
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Designed for particle and cell removal.
  • Bottle top (BT) unit.
  • Disposable filters for reusable receiver flasks and bottles.
  • Optimized for high flow rates and extremely low protein binding and extractables.
  • Well suited for particle removal from tissue culture media, buffers, additives and other aqueous biologicals.
Specifications
Specifications
Clear/Yellow
Hydrophilic
0.45 μm
62 cm2
Bottle Top Filter
For bottles with 45 mm OD
Plug-on Hose Barb
SFCA
Sterile
Polystyrene
500 mL