Learn More
Sartorius Qualitative Grade 292 a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand:  Sartorius FT-3-215-070
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10168531
  • 143.00 SEK / Pack of 100
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
Disc
97 sec.
100/Pk.
70 mm
4 to 7 μm
292 a
0.19 mm