Learn More
Sartorius Qualitative Grade 37/N Filter Papers
This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.
Brand:  Sartorius FT-3-480-150
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11790496
  • 312.00 SEK / Pack of 50
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.
Specifications
Specifications
Disc
37/N
0.5 mm
4 sec.
50/Pk.
150 mm