Learn More
Sartorius Actiosart Syringe Filters
For ultra-cleaning of gases e.g. to absorb unpleasant odors
Brand: Sartorius 17840----------Q
16709.49 SEK valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- MBS housing with high-adsorptive active carbon filling, connector on both sides male Luer slip
- Sample cleaning and particle removal
- Easy to handle
- 100% tested during manufacture
Specifications
Single-use | |
PTFE | |
Syringe Filter |