Learn More
Sartorius Arium™ Comfort Pre-Filter
Product Code. 15744595 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium™ Comfort I and II
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15744595 Arium™ Comfort I and II Each
1 options
Product Code. 15744595 Supplier Sartorius Supplier No. H2OCPFCO1
  • ONLINE EXCLUSIVE PRICE
    4375.00 SEK
    3971.08 SEK / Paket med 1
    Save 403.92 SEK
    9% Av
View all promotions
 Restricted Item
Beräknad leverans: 07-12-2026
Arium™ Comfort Pre-Filter for Arium™ comfort water systems.
  • Pre-filter consisting of high efficient activated carbon combined with a process optimizing catalyst
  • Removes reliable oxidizing agents, heavy metals and particular impurities
  • For intended use with: Arium™ Comfort I Arium™ Comfort II
  • Qty. 1 Unit/Pack
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium™ Comfort I and II
Type Pre-treatment Cartridge