Learn More
Sartorius Entris™ Precision Balances
Delivers effective and reliable weighing results in daily work. Sartorius Entris™ Precision Balances provide easy and clearly structured user interface, logical key assignment and excellent readability for error-free operation.
10726.39 SEK - 18878.45 SEK
Specifications
Readability | 1 mg |
---|---|
Linearity | 2 mg |
Repeatability | 1 mg |
Dimensions (D x W x H) | 230 x 303 x 136 mm |
Diameter (Metric) Outer Pan | 115 mm |
Product Code | Brand | Readability | Model | Stabilization Time | Capacity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Readability | Model | Stabilization Time | Capacity | Price | Quantity & Availability | ||
15263913
|
Sartorius
ENTRIS623-1S |
1 mg | ENTRIS623-1S | 1.0 sec. | 620 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15283913
|
Sartorius
ENTRIS153-1S |
1 mg | ENTRIS153-1S | 1.3 sec. | 150 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15273913
|
Sartorius
ENTRIS423-1S |
1 mg | ENTRIS423-1S | 1.0 sec. | 420 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15293913
|
Sartorius
ENTRIS323-1S |
1 mg | ENTRIS323-1S | 1.1 sec. | 320 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15253903
|
Sartorius
ENTRIS323I-1S |
1 mg | ENTRIS323i-1S | 1.1 sec. | 320 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15223903
|
Sartorius
ENTRIS423I-1S |
1 mg | ENTRIS423i-1S | 1.0 sec. | 420 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15233903
|
Sartorius
ENTRIS153I-1S |
1 mg | ENTRIS153i-1S | 1.3 sec. | 150 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15203903
|
Sartorius
ENTRIS623I-1S |
1 mg | ENTRIS623i-1S | 1.0 sec. | 620 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
- New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
- RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
- Total ease of operation
- All keys have positive click action
- Clearly defined control panel
- Monolithic weighing cell provides high thermal stability and extremely fast response times
- Fast response time and display stability
- Language-driven user interface
- Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Specifications
1 mg | |
1 mg | |
115 mm |
2 mg | |
230 x 303 x 136 mm |