Learn More
Sartorius Funnels for Combisart™
Reusable stainless steel Combisart™ Funnels for microbial enumeration and particle counting.
Brand: Sartorius 6981063
1056.21 SEK valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
- The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
- For traceability, each filter funnel has an individual serial number
Specifications
100 mL | |
Combisart Funnel | |
1 Ea. |
Stainless Steel | |
Small Central Air Port | |
Funnel Lid |