Learn More
Sartorius Lid Seal for Combisart™ Funnels
Lid Seal for use with Sartorius Combisart™ Funnels.
Brand: Sartorius 6981064
129.44 SEK valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
- Lid Seal available in silicone or stainless steel
Specifications
100 mL | |
Seal, Lid | |
Silicone | |
Combisart Funnels |